missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Thrombopoietin R/Tpo R Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF1016
This item is not returnable.
View return policy
Description
Thrombopoietin R/Tpo R Polyclonal specifically detects Thrombopoietin R/Tpo R in Human samples. It is validated for Western Blot, Neutralization.
Specifications
| Thrombopoietin R/Tpo R | |
| Polyclonal | |
| Unconjugated | |
| P40238 | |
| MPL | |
| Mouse myeloma cell line NS0-derived recombinant human Thrombopoietin R/Tpo R Gln26-Tyr423 Accession # P40238 | |
| 100 μg | |
| Primary | |
| Detects human Thrombopoietin R/Tpo R in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human Epo R is observed. | |
| Human | |
| IgG |
| Western Blot, Neutralization | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Neutralization 0.5-2.5 ug/mL | |
| CD110, CD110 antigen, C-MPL, MPL, MPLV, Myeloproliferative leukemia protein, myeloproliferative leukemia virus oncogene, Proto-oncogene c-Mpl, thrombopoietin receptor, TpoR, TPO-R, TPORMPLV | |
| Goat | |
| Antigen Affinity-purified | |
| RUO | |
| 4352 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction