missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Thrombomodulin ELISA Kit

Product Code. 18097404
Change view
Click to view available options
Quantity:
96 Tests
Unit Size:
Pack of 96
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18097404 96 Tests Pack of 96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18097404 Supplier Invitrogen™ Supplier No. EEL020

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Thrombomodulin (TM) is also known as Fetomodulin (FM), endothelial anticoagulant protein, or glycoprotein P112. Thrombomodulin contains six repeated domains homologous with epidermal growth factor (EGF) and an amino terminal domain homologous to lectin-like protein. Through its accelerated activation of protein C, synthesis of TM is one of several mechanisms important in maintaining thrombo-resistance and thus reducing clot formation on the surface of the endothelial cells. Immunohistochemical staining of formalin-fixed tissue for TM has been used for the study of a variety of vascular tumors, meningiomas and choriocarcinomas.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P07204
Assay Range 62.5 to 4000 pg/mL
Assay Sensitivity 37.5 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 7056
Gene Symbol THBD
Interassay CV <10%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Biotinylated Detection Antibody, HRP Conjugate, Standard & Sample Diluent, Biotinylated Detection Antibody Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 100 μL
Storage Requirements Refer to product documentation for component specific storage temperature.
Target Thrombomodulin (CD141)
Test Time 1 hrs 20 mins
Total Assay Time 3 hrs 30 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.