missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Thrombomodulin/BDCA-3 Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB3947-100
This item is not returnable.
View return policy
Description
Thrombomodulin/BDCA-3 Monoclonal specifically detects Thrombomodulin/BDCA-3 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF-ready, ELISA Capture (Matched Antibody Pair).
Specifications
| Thrombomodulin/BDCA-3 | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 8-25 ug/mL, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P07204(Val473) | |
| THBD | |
| Mouse myeloma cell line NS0-derived recombinant human Thrombomodulin/BDCA-3 Ala19-Ser515 Accession # P07204(Val473) | |
| 100 μg | |
| Primary | |
| Detects human Thrombomodulin/BDCA-3 in ELISAs. | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry, CyTOF, ELISA | |
| 501733 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BDCA3, BDCA-3, CD141, CD141 antigen, fetomodulin, THBD, THRM, thrombomodulin, TM | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7056 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction