missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Thioredoxin-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£153.00 - £397.00
Specifications
| Antigen | Thioredoxin-1 |
|---|---|
| Clone | 763629 |
| Dilution | Western Blot 2 ug/mL, Immunocytochemistry 3-25 ug/mL |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18742917
![]() |
R&D Systems
MAB1970-SP |
25 μg |
£153.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18740657
![]() |
R&D Systems
MAB1970-100 |
100 μg |
£397.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Thioredoxin-1 Monoclonal specifically detects Thioredoxin-1 in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
| Thioredoxin-1 | |
| Western Blot 2 ug/mL, Immunocytochemistry 3-25 ug/mL | |
| Monoclonal | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| 7295.0 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
| 763629 | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Human | |
| P10599 | |
| E. coli-derived recombinant human Thioredoxin-1, Met1-Val105, Accession No. P10599 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Thioredoxin-1 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title