Learn More
Human Thimet Oligopeptidase/THOP1 Biotinylated Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand:  R&D Systems BAF3439
Product Code. 18159850
  • £457.00 / 50µg
Description
Thimet Oligopeptidase/THOP1 Polyclonal specifically detects Thimet Oligopeptidase/THOP1 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Thimet Oligopeptidase/THOP1
Polyclonal
Biotin
P52888
THOP1
E. coli-derived recombinant human Thimet Oligopeptidase/THOP1 Lys2-Cys689 Accession # P52888
50 μg
Primary
Detects human Thimet Oligopeptidase/THOP1 in Western blots. In Western blots, approximately 5% cross-reactivity with recombinant human Neurolysin is observed.
Human
IgG
Western Blot
LYOPH
Western Blot 0.1 ug/mL
EC 3.4.24, EC 3.4.24.15, Endopeptidase 24.15, EP24.15, MEPD_HUMAN, MP78, thimet oligopeptidase, thimet oligopeptidase 1, TOP
Sheep
Affinity Purified
RUO
7064
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.