missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TGIF2LX (aa 151-240) Control Fragment Recombinant Protein

Product Code. 30212034
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30212034

Brand: Invitrogen™ RP95452

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (48%), Rat (48%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-56971 (PA5-56971. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

TGIF2LX is a member of the TALE/TGIF homeobox family of transcription factors. Testis-specific expression suggests that this gene may play a role in spermatogenesis. This gene encodes a member of the TALE/TGIF homeobox family of transcription factors. Testis-specific expression suggests that this gene may play a role in spermatogenesis. A homolog of this gene lies within the male specific region of chromosome Y, in a block of sequence that is thought to be the result of a large X-to-Y transposition.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8IUE1
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 90316
Name Human TGIF2LX (aa 151-240) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias homeobox protein TGIF2LX; MGC34726; OTTMUSG00000018465; testis expressed homeobox Te x 1; Te x 1; TGF beta-induced transcription factor 2-like protein; TGFB induced factor homeobox 2 like X-linked; TGFB induced factor homeobox 2 like, X-linked; TGF-beta-induced transcription factor 2-like protein; TGFB-induced factor 2-like protein, X-linked; TGFB-induced factor 2-like, X-linked; TGFB-induced factor homeobox 2-like, X-linked; TGFB-induced factor homeobox 2-like, X-linked 1; TGIF homeobox 1; TGIF2LX; Tgif2l x 1; TGIF-like on the X; TGIFLX; Tgif x 1
Common Name TGIF2LX
Gene Symbol TGIF2LX
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence PDNVQSLPLWPLPKGQMSREKQPDPESAPSQKLTGIAQPKKKVKVSVTSPSSPELVSPEEHADFSSFLLLVDAAVQRAAELELEKKQEPN
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.