missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TGFB1 (P01137) Partial Recombinant Protein expressed in CHO cells

Product Code. 16292460
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
This item is not returnable. View return policy

Product Code. 16292460

Brand: Abnova™ P6032.10ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Used for Func, SDS-PAGE

TGFB is a multifunctional peptide that controls proliferation, differentiation, and other functions in many cell types. TGFB acts synergistically with TGFA (MIM 190170) in inducing transformation. It also acts as a negative autocrine growth factor. Dysregulation of TGFB activation and signaling may result in apoptosis. Many cells synthesize TGFB and almost all of them have specific receptors for this peptide. TGFB1, TGFB2 (MIM 190220), and TGFB3 (MIM 190230) all function through the same receptor signaling systems.[supplied by OMIM]

Sequence: ALDTNYCFSSTEKNCCVRQLYIDFRKDLGWKWIHEPKGYHANFCLGPCPYIWSLDTQYSKVLALYNQHNPGASAAPCCVPQALEPLPIVYYVGRKPKVEQLSNMIVRSCKCS

Specifications

Accession Number P01137
For Use With (Application) Functional Study, SDS-PAGE
Formulation Lyophilized
Gene ID (Entrez) 7040
Molecular Weight (g/mol) 25.0kDa
Name TGFB1 (Human) Recombinant Protein
Preparation Method Mammalian cell (CHO) expression system
Quantity 10 μg
Immunogen ALDTNYCFSSTEKNCCVRQLYIDFRKDLGWKWIHEPKGYHANFCLGPCPYIWSLDTQYSKVLALYNQHNPGASAAPCCVPQALEPLPIVYYVGRKPKVEQLSNMIVRSCKCS
Storage Requirements Store at -20°C. Reconstitute in water to a concentration of 0.1-1.0 mg/mL. Do not vortex. For extended storage, to further dilute in buffer containing carrier protein (e.g. 0.1% BSA) and store in aliquots at -20°C to -80°C is recommended.
Regulatory Status RUO
Endotoxin Concentration <0.1ng/μg (<1 EU/μg)
Gene Alias CED/DPD1/TGFB/TGFbeta
Common Name TGFB1
Gene Symbol TGFB1
Assays Activity assay
SDS-PAGE
The optimal working dilution should be determined by the end user.
Biological Activity Determined by TGFB1's ability to inhibit the mouse IL-4-dependent proliferation of mouse HT-2 cells. The expected ED50 is ≤ 0.05ng/mL, corresponding to a specific activity of >= 2 x 107 units/mg.
Cross Reactivity Bovine,Chicken,Dog,Frog,Monkey,Mouse,Pig,Rabbit,Rat ;Bovine,Chicken,Dog,Frog,Monkey,Mouse,Pig,Rabbit,Rat
Species Human
Recombinant Recombinant
Protein Tag None
Expression System Mammalian cell (CHO) expression system
Form Lyophilized
Purity or Quality Grade 0.98
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.