missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TGF-beta RII Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems BAF241
This item is not returnable.
View return policy
Description
TGF-beta RII Polyclonal specifically detects TGF-beta RII in Human samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).
Specifications
| TGF-beta RII | |
| Polyclonal | |
| Biotin | |
| P37173 | |
| TGFBR2 | |
| Mouse myeloma cell line NS0-derived recombinant human TGF-beta RII Ile24-Asp159 Accession # P37173 | |
| 50 μg | |
| Primary | |
| Detects human TGF-beta RII in ELISAs and Western blots. In sandwich immunoassays, approximately 25% cross-reactivity with recombinant human (rh) TGF-beta RIIb is observed and less than 0.05% cross-reactivity with rhTGF-beta RI, rhTGF-beta RIII, recombinant mouse TGF-beta RII, and rhTGF-beta 1 is observed. | |
| Human | |
| IgG |
| Western Blot, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
| AAT3, EC 2.7.11, EC 2.7.11.30, FAA3, HNPCC6, LDS1B, LDS2B, MFS2, RIIC, TAAD2, tbetaR-II, TGF-beta receptor type II, TGF-beta receptor type IIB, TGF-beta receptor type-2, TGF-beta type II receptor, TGFbetaRII, TGFbeta-RII, TGFBR2, TGF-bRII, TGFR-2, transforming growth factor beta receptor type IIC, transforming growth factor, beta receptor II (70/80kDa), transforming growth factor, beta receptor II (70/80kDa) isoform 1, transforming growth factor, beta receptor II (70/80kDa) isoform 2, transforming growth factor, beta receptor II (70-80kD), Transforming growth factor-beta receptor type II | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 7048 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction