missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TGF-beta RI/ALK-5 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB3025-100
This item is not returnable.
View return policy
Description
TGF-beta RI/ALK-5 Monoclonal specifically detects TGF-beta RI/ALK-5 in Human samples. It is validated for ELISA.
Specifications
| TGF-betaRI/ALK-5 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
| AAT5, activin A receptor type II-like kinase, 53kD, activin A receptor type II-like kinase, 53kDa, Activin receptor-like kinase 5, ACVRLK4, ALK-5, ALK-5ALK5, EC 2.7.11, EC 2.7.11.30, LDS1A, LDS2A, Serine/threonine-protein kinase receptor R4, SKR4, TbetaR-I, TGFB1R1, TGF-beta receptor type I, TGF-beta receptor type-1, TGF-beta type I receptor, TGFbetaRI, TGFBR1, TGF-bRI, TGFR-1, transforming growth factor beta receptor I, transforming growth factor, beta receptor 1, transforming growth factor, beta receptor I (activin A receptor type II-likekinase, 53kD), Transforming growth factor-beta receptor type I | |
| Mouse myeloma cell line NS0-derived recombinant human TGF-beta RI/ALK-5 Leu34-Glu125 Accession # P36897 | |
| 100 μg | |
| 7046 | |
| Reconstitute at 0.5mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| ELISA | |
| 854621R | |
| ELISA | |
| P36897 | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects human TGF-beta RI/ALK-5 in direct ELISAs. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction