missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human TGF-beta 3 Recombinant Protein
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Description
Recombinant human TGF-beta3 is a non-glycosylated, disulfide-linked homodimer, containing two 112 amino acid chains.
Transforming growth factor beta 3 (TGF beta 3) is the third member of the transforming growth factor family of cytokines, which also includes TGF beta 1 and beta 2. These cytokines are secreted in precursor form consisting of a bioactive C-terminal domain attached to an N-terminal domain known as latency associated protein (LAP). Cleavage of LAP results in the mature protein, which functions as a disulfide-linked homodimer. As with all members of the family, TGF beta 3 is highly conserved across species, with mouse and human TGF beta 3 demonstrating 100% sequence homology and cross-species activity.The members of this family can be expressed by most cell types and exert pleiotropic effects, which include the suppression of B- and T-cell effector activity, mediation of tissue healing, and suppression of tumor proliferation. The promotion of CD4+CD25+ T-cell expansion is a newly discovered function of the TGF beta cytokines, and indicates an important role in the protection against autoimmunity.
Specifications
Specifications
| Accession Number | P10600 |
| Concentration | 0.25 mg/mL |
| For Use With (Application) | Control |
| Formulation | 0.01 M acetic acid with 20% ethanol and no preservative |
| Gene ID (Entrez) | 7043 |
| Molecular Weight (g/mol) | 12.9 kDa |
| Name | Human TGF-beta 3 |
| Quantity | 10 μg |
| Source | E. coli |
| Regulatory Status | RUO |
| Show More |
Safety and Handling
Product Identifier
- Human TGF-beta 3 Recombinant Protein
- Warning
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P240-Ground/bond container and receiving equipment.
- P243-Take precautionary measures against static discharge.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : ethanol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction