missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human TGF-beta 2 Recombinant Protein, PeproTech®

Product Code. 17859992
Click to view available options
Quantity:
10 μg
100 μg
2 μg
2 x 250 μg
250 μg
4 x 250 μg
Unit Size:
100µg
10µg
1mg
250µg
2µg
500µg
This item is not returnable. View return policy

Product Code. 17859992

Brand: Gibco™ 10035250UG

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Insect cells Recombinant Protein

Transforming Growth Factor (TGF) betas mediate many cell to cell interactions that occur during embryonic development. Three TGF betas have been identified in mammals. TGF beta 1, TGF beta 2 and TGF beta 3 are each synthesized as precursor proteins that are very similar in that each is cleaved to yield a 112 amino acid polypeptide that remains associated with the latent portion of the molecule. The TGF beta polypeptides are multifunctional; capable of influencing cell proliferation, differentiation, and other functions in a wide range of cell types. Transformed, as well as nonneoplastic tissues, release transforming growth factors; and essentially all mammalian cells possess a specific TGF receptor. The multi modal nature of TGF beta is seen in its ability to stimulate or inhibit cellular proliferation. In general, cells of mesenchymal origin appear to be stimulated by TGF beta whereas cells of epithelial or neuroectodermal origin are inhibited by the peptide. TGF beta 1, TGF beta 2, and TGF beta 1.2 appear to be equivalent in biological activity, although there does appear to be differences in binding to certain types of receptors. TGF beta 2 is produced by many cell types and has been found in the highest concentration in porcine platelets and mammalian bone. Latent TGF beta 2 is the prominent isoform found in body fluids such as amniotic fluid, breast milk, and the aqueous and vitreous humor of the eye.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P61812
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 7042
Molecular Weight (g/mol) 25
Name Human TGF-beta 2
Quantity 250 μg
Source Insect cells
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias BB105277; BSC-1 cell growth inhibitor; cetermin; glioblastoma-derived T-cell suppressor factor; G-TSF; LAP; Latency-associated peptide; LDS4; polyergin; prepro-transforming growth factor beta-2; TGF beta 2 protein; TGFB; TGFB2; TGF-B2; Tgfb-2; TGF-beta2; TGF-beta-2; TGFß2; Transforming growth factor; transforming growth factor beta 2; transforming growth factor beta-2; Transforming growth factor beta-2 proprotein; transforming growth factor, beta 2
Gene Symbol TGFB2
Specific Reactivity Human
Conjugate Unconjugated
Recombinant Recombinant
Sequence ALDAAYCFRN VQDNCCLRPL YIDFKRDLGW KWIHEPKGYN ANFCAGACPY LWSSDTQHSR VLSLYNTINP EASASPCCVS QDLEPLTILY YIGKTPKIEQ LSNMIVKSCK CS
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.