missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ Human TGF-beta 2 Platinum ELISA Kit
Click to view available options
Quantity:
96 Tests
Unit Size:
96 tests
Description
Includes
- Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to human TGF-beta2
- Biotin-Conjugate anti-human TGF-beta2 monoclonal antibody
- Streptavidin-HRP
- Human TGF-beta2 Standard, 1ng/mL
- Assay Buffer Concentrate 10x
- Wash Buffer Concentrate 40x
- Substrate Solution (tetramethyl-benzidine)
- Stop Solution (0.5M H2SO4)
- Adhesive Films
Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications
Specifications
| Assay Sensitivity | 6.6pg/mL |
| Conjugate | Biotin |
| Sample Type | Cell Culture Supernatant, Serum |
| Target Species | Human |
| For Use With (Application) | ELISA |
| Quantity | 96 Tests |
| Sample Volume | 100 μL (1:50 prediluted) |
| Synonym | G-TSF, Polyergin, Cetermin, BSC-1 |
| Target | TGF beta 2 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction