missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human/Mouse/Rat/Porcine/Canine TGF-beta 1 Quantikine ELISA
Human TGF-beta 1 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Specifications
| Assay Range | 31.20 - 2000pg/mL |
|---|---|
| Assay Sensitivity | 15.4pg/mL |
| Sample Type | Cell Culture Supernates (100 uL), Serum (40 uL), EDTA Plasma (40 uL), Platelet-poor Plasma (25 uL), Urine (100 uL), Plasma (EDTA), Serum, Urine |
Description
The Human TGF-beta 1 Quantikine ELISA Kit quantifies human TGF-beta 1 in cell culture supernates (100 ul), serum (40 ul), edta plasma (40 ul), urine (100 ul).Specifications
| 31.20 - 2000pg/mL | |
| 15.4pg/mL | |
| Cell Culture Supernates (100 uL), Serum (40 uL), EDTA Plasma (40 uL), Platelet-poor Plasma (25 uL), Urine (100 uL), Plasma (EDTA), Serum, Urine |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title