missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human TGF-alpha Recombinant Protein, PeproTech®

Product Code. 17850953
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
500 μg
Unit Size:
100µg
1mg
20µg
250µg
500µg
This item is not returnable. View return policy

Product Code. 17850953

Brand: Gibco™ 10016A250UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Recombinant human TGF-alpha is a 50 amino acid polypeptide (5.5 kDa) which shares approximately 40% sequence homology with EGF, including 6 conserved cysteine residues, which form 3 intramolecular disulfide bonds. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

TGF-alpha is an EGF-related polypeptide growth factor that signals through the EGF receptor, and stimulates the proliferation of a wide range of epidermal and epithelial cells. It is produced by monocytes, keratinocytes, and various tumor cells. TGF-alpha induces anchorage-independence transformation in cultured cells. Human, murine and rat TGF-alpha are cross-species reactive. TGFalpha (aa 50) is a growth factor with 33% homology to EGF, binds to EGFR, activates tyrosine phosphorylation of the receptor, and stimulates cell proliferation. It plays a role in tumor initiation by inducing the reversible transformed phenotype.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01135
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 7039
Molecular Weight (g/mol) 5.5 kDa
Name Human TGF-alpha
Quantity 250 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias EGF-like TGF; ETGF; I79_019067; LOC100009150; Protransforming growth factor alpha; RATTGFAA; TFGA; TGF a; TGF type 1; TGF a; TGFA; TGFAA; TGFalpha; TGF-alpha; TGFa; Transforming growth factor; Transforming growth factor alpha; transforming growth factor, alpha; transforming growth factor-alpha; wa1; wa-1; waved 1
Common Name TGF alpha
Gene Symbol TGFA
Biological Activity The ED50 as determined by the dose-dependent stimulation of thymidine uptake by BALB/c 3T3 cells is ≤ 0.2 ng/ml, corresponding to a specific activity of ≥ 5 x 10^6 units/mg
Conjugate Unconjugated
Recombinant Recombinant
Sequence VVSHFNDCPD SHTQFCFHGT CRFLVQEDKP ACVCHSGYVG ARCEHADLLA
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.