missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Human TGF-alpha Recombinant Protein

Product Code. 18610254 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18610254 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18610254 Supplier Novus Biologicals™ Supplier No. NBP236432100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

The ED50, as determined by a cell proliferation assay using murine Balb/3T3 cells, is less than 0.4 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) SDS-PAGE, Bioactivity
Gene ID (Entrez) 7039
Molecular Weight (g/mol) MolecularWeight-theroretical: 6 kDa
Quantity 100 μg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Endotoxin Concentration Less than 1 EU/mg of the rHuTGF-a as determined by LAL method.
Gene Alias protransforming growth factor alpha, TFGA, transforming growth factor, alpha, transforming growth factor-alpha
Gene Symbol TGFA
Reconstitution Reconstitute with 10 mM acetic acid to a final concentration of 0.1 mg/mL
Research Category Angiogenesis, Cell Cycle and Replication, Signal Transduction
Purity or Quality Grade SDS-PAGE
Protein TGF-alpha
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.