missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TG native Protein from human thyroid

Product Code. 16242270
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy

Product Code. 16242270

Brand: Abnova™ P5343.5mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for SDS-PAGE

Specifications

For Use With (Application) SDS-PAGE
Formulation Lyophilized from 0.02M ammonium bicarbonate andMay contain traces of buffer salts
Gene ID (Entrez) 7038
Name TG (Human) Native Protein
Preparation Method Native protein purified from human thyroid
Quantity 5 mg
Storage Requirements Store at 4°C. This product is stable for at least 3 months. After reconstitution with phosphate buffer, pH 7.0 (contains 0.15 M NaCl), aliquot and store at 4°C.
Regulatory Status RUO
Gene Alias AITD3/TGN
Common Name TG
Gene Symbol TG
Species Human
Protein Tag None
Expression System Native protein purified from human thyroid
Form Lyophilized
Purity or Quality Grade ≥96%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.