missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TFPI Biotinylated Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems BAF2974
This item is not returnable.
View return policy
Description
TFPI Polyclonal specifically detects TFPI in Human samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).
Specifications
| TFPI | |
| Polyclonal | |
| Biotin | |
| P10646 | |
| TFPI | |
| Mouse myeloma cell line NS0-derived recombinant human TFPI Asp29-Lys282 Accession # P10646 | |
| 50 μg | |
| Primary | |
| Detects human TFPI in ELISAs and Western blots. In sandwich immunoassays, less than 0.04% cross-reactivity with recombinant mouse (rm) TFPI, recombinant human TFPI-2, and rmTFPI-2 is observed. | |
| Human | |
| IgG |
| Western Blot, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
| convertin, EPI, EPITFPI1LACITFI, Extrinsic pathway inhibitor, LACI, Lipoprotein-associated coagulation inhibitor, tissue factor pathway inhibitor, tissue factor pathway inhibitor (lipoprotein-associated coagulation inhibitor) | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 7035 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction