missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TFPI Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 3 publications
Brand: R&D Systems AF2974
This item is not returnable.
View return policy
Description
TFPI Polyclonal specifically detects TFPI in Human samples. It is validated for Western Blot, Simple Western, Immunoprecipitation.
Specifications
| TFPI | |
| Polyclonal | |
| Unconjugated | |
| P10646 | |
| TFPI | |
| Mouse myeloma cell line NS0-derived recombinant human TFPI Asp29-Lys282 Accession # P10646 | |
| 100 μg | |
| Primary | |
| Detects human TFPI in direct ELISAs and Western blots. In directs ELISAs, less than 10% cross-reactivity with recombinant mouse (rm) TFPI is observed and less than 1% cross-reactivity with recombinant human TFPI-2, and rmTFPI-2 is observed. | |
| Human | |
| IgG |
| Western Blot, Immunoprecipitation | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 50 ug/mL, Immunoprecipitation 25 ug/mL | |
| convertin, EPI, EPITFPI1LACITFI, Extrinsic pathway inhibitor, LACI, Lipoprotein-associated coagulation inhibitor, tissue factor pathway inhibitor, tissue factor pathway inhibitor (lipoprotein-associated coagulation inhibitor) | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 7035 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction