missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Tenascin X Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB6999
This item is not returnable.
View return policy
Description
Tenascin X Monoclonal specifically detects Tenascin X in Human samples. It is validated for Western Blot, Simple Western.
Specifications
| Tenascin X | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL | |
| AAH33740 | |
| TNXB | |
| Chinese hamster ovary cell line CHO-derived recombinant human Tenascin XB2 Met1-Tyr673 Accession # AAH33740 | |
| 100 μg | |
| Primary | |
| Detects human Tenascin XB2 in ELISAs and Western blots. Also detects Tenascin X in Western blots. In direct ELISAs, no cross-reactivity with recombinant human Tenascin R is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 766842 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Hexabrachion-like protein, HXBL, tenascin XB, tenascin XB2, tenascin-X, TNX, TNXB, TNXB1growth-inhibiting protein 45, TNXB2, TNXB2tenascin XB1, TNXBS, TNXBSTNXTENX, XB, XBS, XB-S, XBTN-X | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7148 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto