missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Tenascin C ELISA Kit

Product Code. 17175229
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 96
This item is not returnable. View return policy

Product Code. 17175229

Brand: Invitrogen™ EH446RBX5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Human Tenascin C quantitates human Tenascin C in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Tenascin C.

Tenascin is a high molecular weight, multifunctional, extracellular matrix glycoprotein expressed in association with mesenchymal-epithelial interactions during development and in the neovasculature and stroma of undifferentiated tumors (Erickson and Bourdon 1989). It has been described under a variety of names: cytotactin, hexabrachion protein, J1, myotendinous antigen (MI) and glioma mesenchymal extracellular matrix (GMEM). The tenascin molecule is a disulfide-linked hexamer. The expression of tenascin is associated with development and growth, both normal and pathological, whereas the distribution in normal adult tissue is restricted.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number 150-225; AI528729; C130033P17Rik; cytotactin; deafness, autosomal dominant 56; DFNA56; Glioma-associated-extracellular matrix antigen; GMEM; GP; GP 150-225; hexabrachion; hexabrachion (tenascin); Hxb; JI; MGC144208; MGC144209; MGC167029; Myotendinous antigen; neuronectin; Ten; tenascin; tenascin C; Tenascin-C; tenascin-C additional domain 1; tenascin-C isoform 14/AD1/16; TN; Tnc; TN-C
Assay Range 6.1 to 1,500 pg/mL
Assay Sensitivity 6.1 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 3371
Gene Symbol TNC
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target Tenascin C
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.