missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TEM5 (aa 158-271) Control Fragment Recombinant Protein

Product Code. 30210030
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30210030

Brand: Invitrogen™ RP109719

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (89%), Rat (89%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-144689 (PA5-144689. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Tumor endothelial markers (TEMs) are significantly up-regulated during angiogenesis and neoangiogensis that are crucial for the growth of solid tumors. TEMs localized on the cell surface and conserved across species are of particular interest for future development of anti-angiogenic therapies. These include TEMs such as TEM1, TEM5, TEM7 and TEM8. TEM5 is a member of the adhesion family of G protein coupled receptors and is localized on the surface of endothelial cells. TEM5 is a seven-pass transmembrane receptor, unlike TEM1, TEM7 and TEM8 which span the membrane once. TEM5 is abundantly expressed in tumor vessels, heart, placenta, ovary, small intestine, and colon. Proteolytically processed soluble TEM5 mediates endothelial cell survival during angiogenesis by linking integrin to glycosaminoglycans.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q96PE1
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 25960
Name Human TEM5 (aa 158-271) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 8430414O08Rik; 9530074E10Rik; Adgra2; Adgra3; Adhesion G protein-coupled receptor A2; adhesion G protein-coupled receptor A3; DKFZp434C211; DKFZp434J0911; FLJ14390; G protein-coupled receptor 124; Gpr124; G-protein coupled receptor 124; KIAA1531; LOC100363275; mKIAA1531; Tem5; Tumor endothelial marker 5
Common Name TEM5
Gene Symbol ADGRA2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LLRLNISGNIFSSLQPGVFDELPALKVVDLGTEFLTCDCHLRWLLPWAQNRSLQLSEHTLCAYPSALHAQALGSLQEAQLCCEGALELHTHHLIPSLRQVVFQGDRLPFQCSAS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.