missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human TECPR2 (aa 1270-1411) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Beskrivning
Highest antigen sequence indentity to the following orthologs: Mouse (89%), Rat (89%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-51435 (PA5-51435. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Specifikationer
Specifikationer
| Accession Number | O15040 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 9895 |
| Name | Human TECPR2 (aa 1270-1411) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | KIAA0297; KIAA0329; SPG49; TECPR2; tectonin beta-propeller repeat containing 2; tectonin beta-propeller repeat-containing protein 2; WD repeat-containing protein KIAA0329/KIAA0297 |
| Common Name | TECPR2 |
| Visa mer |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering