missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TEAD4 (aa 139-202) Control Fragment Recombinant Protein

Product Code. 30204370
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30204370

Brand: Invitrogen™ RP101475

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (89%), Rat (89%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene product is a member of the transcriptional enhancer factor (TEF) family of transcription factors, which contain the TEA/ATTS DNA-binding domain. It is preferentially expressed in the skeletal muscle, and binds to the M-CAT regulatory element found in promoters of muscle-specific genes to direct their gene expression. Alternatively spliced transcripts encoding distinct isoforms, some of which are translated through the use of a non-AUG (UUG) initiation codon, have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q15561
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 7004
Name Human TEAD4 (aa 139-202) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias EFTR-2; Etfr2; ETFR-2; ETF-related factor 2; ETF-related factor-2; FGF-regulated 19; FR-19; hRTEF-1 B; related transcription enhancer factor 1 B; RTEF1; RTEF-1; TCF13L1; Tcf13r1; TEA domain family member 4; TEA domain transcription factor 4; TEAD4; TEAD-4; TEF-1-related factor 1; TEF-1-related factor FR-19; TEF3; TEF-3; Tefr; Tefr1; TEFR-1; Tefr1a; Transcription factor 13-like 1; transcription factor RTEF-1; transcriptional enhancer factor 1-related; transcriptional enhancer factor 3; transcriptional enhancer factor TEF-3
Common Name TEAD4
Gene Symbol Tead4
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence ISATAFHSSMALARGPGRPAVSGFWQGALPGQAGTSHDVKPFSQQTYAVQPPLPLPGFESPAGP
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.