missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TCF7L1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB61161
This item is not returnable.
View return policy
Description
TCF7L1/TCF3 Monoclonal specifically detects TCF7L1/TCF3 in Human samples. It is validated for Immunohistochemistry.
Specifications
| TCF7L1/TCF3 | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 8-25 ug/mL | |
| Q9HCS4 | |
| TCF7L1 | |
| E.coli-derived recombinant human TCF7L1 Lys429-Ser581 Accession # Q9HCS4 | |
| 100 μg | |
| Primary | |
| Detects human TCF7L1 in direct ELISAs. In direct ELISAs, 5-10% cross-reactivity with recombinant human (rh) TCF-4 (aa 102-516) and rhTCF-12 is observed. No cross-reactivity with rhTCF7L1 (aa 151-320), rhTCF7L2, rhTCF-1, -2, -3, -4 (aa 241-365), rhLEF-1, or recombinant mouse TCF-1 is observed. | |
| Human | |
| Purified |
| Immunohistochemistry | |
| 622782 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| HMG box transcription factor 3, TCF3, TCF-3, transcription factor 7-like 1, transcription factor 7-like 1 (T-cell specific, HMG-box) | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 83439 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction