missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TCF20 (aa 501-609) Control Fragment Recombinant Protein

Product Code. 30198141
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30198141

Brand: Invitrogen™ RP95352

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (89%), Rat (89%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-57816 (PA5-57816. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a transcription factor that recognizes the platelet-derived growth factor-responsive element in the matrix metalloproteinase 3 promoter. The encoded protein is thought to be a transcriptional coactivator, enhancing the activity of transcription factors such as JUN and SP1. Mutations in this gene are associated with autism spectrum disorders. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifikationer

Accession Number Q9UGU0
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 6942
Name Human TCF20 (aa 501-609) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2810438H08Rik; AR1; KIAA0292; mKIAA0292; Nuclear factor SPBP; Protein AR1; SPBP; SPRE-binding protein; stromelysin 1 PDGF responsive element binding protein; stromelysin 1 PDGF-responsive element-binding protein; stromelysin PDGF responsive element binding protein transcription factor; stromelysin-1 PDGF-responsive element-binding protein; stromelysin-1 platelet-derived growth factor-responsive element binding protein; Tcf20; TCF-20; Transcription factor 20; transcription factor 20 (AR1)
Common Name TCF20
Gene Symbol TCF20
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence SEGSSQPEEQLKSPMAESLDGGCSSSSEDQGERVRQLSGQSTSSDTTYKGGASEKAGSSPAQGAQNEPPRLNASPAAREEATSPGAKDMPLSSDGNPKVNEKTVGVIVS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Visa mer Visa mindre
Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.