missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human TBX2 Partial ORF (NP_005985, 603 a.a. - 702 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00006909-Q01.10ug
This item is not returnable.
View return policy
Description
This gene is a member of a phylogenetically conserved family of genes that share a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. This gene product is the human homolog of mouse Tbx2, and shares strong sequence similarity with Drosophila omb protein. Expression studies indicate that this gene may have a potential role in tumorigenesis as an immortalizing agent. Transcript heterogeneity due to alternative polyadenylation has been noted for this gene. [provided by RefSeq]
Sequence: GSARPRLRFSPYQIPVTIPPSTSLLTTGLASEGSKAAGGNSREPSPLPELALRKVGAPSRGALSPSGSAKEAANELQSIQRLVSGLESQRALSPGRESPKSpecifications
NP_005985 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
GSARPRLRFSPYQIPVTIPPSTSLLTTGLASEGSKAAGGNSREPSPLPELALRKVGAPSRGALSPSGSAKEAANELQSIQRLVSGLESQRALSPGRESPK | |
RUO | |
TBX2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
6909 | |
TBX2 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ10169 | |
TBX2 | |
Recombinant | |
wheat germ expression system |