missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TBX2 Partial ORF (NP_005985, 603 a.a. - 702 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00006909-Q01.25ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16014805

  • £425.00 / 25µg
Estimated Shipment: 06-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene is a member of a phylogenetically conserved family of genes that share a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. This gene product is the human homolog of mouse Tbx2, and shares strong sequence similarity with Drosophila omb protein. Expression studies indicate that this gene may have a potential role in tumorigenesis as an immortalizing agent. Transcript heterogeneity due to alternative polyadenylation has been noted for this gene. [provided by RefSeq]

Sequence: GSARPRLRFSPYQIPVTIPPSTSLLTTGLASEGSKAAGGNSREPSPLPELALRKVGAPSRGALSPSGSAKEAANELQSIQRLVSGLESQRALSPGRESPK
Specifications

Specifications

NP_005985
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
GSARPRLRFSPYQIPVTIPPSTSLLTTGLASEGSKAAGGNSREPSPLPELALRKVGAPSRGALSPSGSAKEAANELQSIQRLVSGLESQRALSPGRESPK
RUO
TBX2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
6909
TBX2 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ10169
TBX2
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers