missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TBX2 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB5040-SP
This item is not returnable.
View return policy
Description
TBX2 Monoclonal specifically detects TBX2 in Human samples. It is validated for Western Blot.
Specifications
| TBX2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| Q13207 | |
| TBX2 | |
| E. coli-derived recombinant human TBX2 Thr593-Arg702 Accession # Q13207 | |
| 25 μg | |
| Primary | |
| Detects endogenous human TBX2 in Western blots at approximately 75 kDa. | |
| Human | |
| Purified |
| Western Blot | |
| 393725 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FLJ10169, T-box 2, T-box protein 2, T-box transcription factor TBX2 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6909 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction