missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TBX18 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB63371-SP
This item is not returnable.
View return policy
Description
TBX18 Monoclonal specifically detects TBX18 in Human samples. It is validated for Immunocytochemistry.
Specifications
| TBX18 | |
| Monoclonal | |
| LYOPH | |
| Immunocytochemistry 8-25 ug/mL | |
| O95935 | |
| TBX18 | |
| E. coli-derived recombinant human TBX18 Ala384-Ser504 Accession # O95935 | |
| 25 μg | |
| Primary | |
| Detects human TBX18 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) TBX2, 3, 5, 6, 20, or rhT-bet is observed. | |
| Human | |
| Purified |
| Immunocytochemistry | |
| 635305 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| T-box 18, T-box protein 18, T-box transcription factor TBX18 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 9096 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction