missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Tapasin (aa 265-388) Control Fragment Recombinant Protein

Product Code. 30210459
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30210459

Brand: Invitrogen™ RP90110

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (73%), Rat (73%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-52382 (PA5-52382. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a transmembrane glycoprotein which mediates interaction between newly assembled major histocompatibility complex (MHC) class I molecules and the transporter associated with antigen processing (TAP), which is required for the transport of antigenic peptides across the endoplasmic reticulum membrane. This interaction is essential for optimal peptide loading on the MHC class I molecule. Up to four complexes of MHC class I and this protein may be bound to a single TAP molecule. This protein contains a C-terminal double-lysine motif (KKKAE) known to maintain membrane proteins in the endoplasmic reticulum. This gene lies within the major histocompatibility complex on chromosome 6. Alternative splicing results in three transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O15533
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 6892
Name Human Tapasin (aa 265-388) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias D17Wsu91e; DADB-159G18.7; NGS17; NGS-17; similar to Tapasin precursor (TPSN) (TPN) (TAP-binding protein) (TAP-associated protein) (NGS-17); TAP binding protein; TAP binding protein (tapasin); TAPA; tapasin; TAP-associated protein; TAP-binding protein; Tapbp; TPN; TPSN
Common Name Tapasin
Gene Symbol TAPBP
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence QEGTYLATIHLPYLQGQVTLELAVYKPPKVSLMPATLARAAPGEAPPELLCLVSHFYPSGGLEVEWELRGGPGGRSQKAEGQRWLSALRHHSDGSVSLSGHLQPPPVTTEQHGARYACRIHHPS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.