missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Tankyrase (aa 80-145) Control Fragment Recombinant Protein

Product Code. 30213369
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30213369

Brand: Invitrogen™ RP93906

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (62%), Rat (62%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Poly-ADP-ribosyltransferase involved in various processes such as Wnt signaling pathway, telomere length and vesicle trafficking (PubMed:10988299, PubMed:11739745, PubMed:16076287, PubMed:19759537, PubMed:21478859, PubMed:22864114, PubMed:23622245, PubMed:25043379). Acts as an activator of the Wnt signaling pathway by mediating poly-ADP-ribosylation (PARsylation) of AXIN1 and AXIN2, 2 key components of the beta-catenin destruction complex: poly-ADP-ribosylated target proteins are recognized by RNF146, which mediates their ubiquitination and subsequent degradation (PubMed:19759537, PubMed:21478859). Also mediates PARsylation of BLZF1 and CASC3, followed by recruitment of RNF146 and subsequent ubiquitination (PubMed:21478859). Mediates PARsylation of TERF1, thereby contributing to the regulation of telomere length (PubMed:11739745). Involved in centrosome maturation during prometaphase by mediating PARsylation of HEPACAM2/MIKI (PubMed:22864114). May also regulate vesicle trafficking and modulate the subcellular distribution of SLC2A4/GLUT4-vesicles (PubMed:10988299). May be involved in spindle pole assembly through PARsylation of NUMA1 (PubMed:16076287). Stimulates 26S proteasome activity (PubMed:23622245). [UniProt]
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O95271
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 8658
Name Human Tankyrase (aa 80-145) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 4930554K12Rik; ADP-ribosyltransferase diphtheria toxin-like 5; AI662855; ARTD5; C86528; D130072O21Rik; HGNC:11941; LOW QUALITY PROTEIN: tankyrase-1; mTNKS1; PARP5A; PARP-5 A; PARPL; pART5; Poly [ADP-ribose] polymerase 5 A; Poly [ADP-ribose] polymerase tankyrase-1; poly [ADP-ribose] polymerase tankyrase-1; tankyrase-1; Protein poly-ADP-ribosyltransferase tankyrase-1; TANK1; tankyrase; tankyrase 1; tankyrase I; tankyrase, TRF1-interacting ankyrin-related ADP-ribose polymerase; tankyrase-1; TIN1; TINF1; TNKS; TNKS1; TNKS-1; TRF1-interacting ankyrin-related ADP-ribose polymerase; TRF1-interacting ankyrin-related ADP-ribose polymerase 1
Common Name Tankyrase
Gene Symbol TNKS
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence SPDPVDGTSCCSTTSTICTVAAAPVVPAVSTSSAAGVAPNPAGSGSNNSPSSSSSPTSSSSSSPSS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.