missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human TAIP2 (aa 346-427) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (93%), Rat (93%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53664 (PA5-53664. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
TAIP2 binds to the consensus sequence 5'-AGAGTG-3' and has transcriptional activator activity. Plays a role in apoptosis.
Specifications
Specifications
| Accession Number | Q8WYN3 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 80034 |
| Name | Human TAIP2 (aa 346-427) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | A330102K23Rik; Csnrp3; Csrnp3; CSRNP-3; cysteine and serine rich nuclear protein 3; cysteine/serine-rich nuclear protein 3; cysteine-serine-rich nuclear protein 3; Fam130a2; family with sequence similarity 130, member A2; Mbu1; PPP1R73; Protein FAM130A2; protein phosphatase 1, regulatory subunit 73; RGD1306214; TAIP2; TAIP-2; TGF beta induced apotosis protein 2; TGF-beta induced apoptosis protein 2; TGF-beta-induced apoptosis protein 2 |
| Common Name | TAIP2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction