missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TAFA5/FAM19A5 ELISA Kit

Product Code. 17165229
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 96
This item is not returnable. View return policy

Product Code. 17165229

Brand: Invitrogen™ EH445RBX5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Human TAFA5/FAM19A5 quantitates human TAFA5/FAM19A5 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human TAFA5/FAM19A5.

Acts as a chemokine-like protein by regulating cell proliferation and migration through activation of G protein-coupled receptors (GPCRs), such as S1PR2 and FPR2. Stimulates chemotactic migration of macrophages mediated by the MAPK3/ERK1 and AKT1 pathway. Blocks TNFSF11/RANKL-induced osteoclast formation from macrophages by inhibiting up-regulation of osteoclast fusogenic and differentiation genes. Stimulation of macrophage migration and inhibition of osteoclast formation is mediated via GPCR FPR2. Acts as an adipokine by negatively regulating vascular smooth muscle cell (VSMC) proliferation and migration in response to platelet-derived growth factor stimulation via GPCR S1PR2 and G protein GNA12/GNA13-transmitted RHOA signaling . Inhibits injury-induced cell proliferation and neointima formation in the femoral arteries
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Chemokine-like protein TAFA-5; Fam19a5; family with sequence similarity 19 (chemokine (C-C motif)-like), member A5; family with sequence similarity 19 member A5, C-C motif chemokine like; family with sequence similarity 19, member A5; protein FAM19A5; QLLK5208; RGD1562115; TAFA protein 5; TAFA5; Tafa-5; Tara-5; UNQ5208; UNQ5208/PRO34524
Assay Range 15 to 4,000 pg/mL
Assay Sensitivity 15 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 25817
Gene Symbol TAFA5
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target TAFA5
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.