missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TAF6 (aa 114-251) Control Fragment Recombinant Protein

Product Code. 30205151
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30205151

Brand: Invitrogen™ RP89319

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (98%), Rat (98%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-52310 (PA5-52310. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Initiation of transcription by RNA polymerase II requires the activities of more than 70 polypeptides. The protein that coordinates these activities is transcription factor IID (TFIID), which binds to the core promoter to position the polymerase properly, serves as the scaffold for assembly of the remainder of the transcription complex, and acts as a channel for regulatory signals. TFIID is composed of the TATA-binding protein (TBP) and a group of evolutionarily conserved proteins known as TBP-associated factors or TAFs. TAFs may participate in basal transcription, serve as coactivators, function in promoter recognition or modify general transcription factors (GTFs) to facilitate complex assembly and transcription initiation. This gene encodes one of the smaller subunits of TFIID that binds weakly to TBP but strongly to TAF1, the largest subunit of TFIID. Four isoforms have been identified but complete transcripts have been determined for only three isoforms. One of the isoforms has been shown to preclude binding of one of the other TFIID subunits, thereby reducing transcription and initiating signals that trigger apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P49848
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 6878
Name Human TAF6 (aa 114-251) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 80 kDa; AW549759; DKFZp781E21155; MGC:8964; p80; RNA polymerase II TBP-associated factor subunit E; RNA polymerase II, TATA box binding protein (TBP)-associated factor, 80 kDa; TAF(II)70; TAF(II)80; TAF2E; Taf6; TAF6 RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF6 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 80 kDa; TAF6 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 80 kDa; TAFII70; TAFII-70; TAFII80; TAFII-80; TAFII85; TATA box binding protein (Tbp)-associated factor, RNA polymerase II, E; TATA box binding protein (TBP)-associated factor, RNA polymerase II, E, 70/85 kD; TATA box binding protein associated factor 6; TATA-box binding protein associated factor 6; TBP-associated factor 6; TBP-associated factor 6 isoform alpha; TFIID subunit p80; transcription initiation factor TFIID 70 kDa subunit; Transcription initiation factor TFIID 80 kDa subunit; transcription initiation factor TFIID subunit 6; wu:fb83e08; wu:fj30b02; zgc:92160
Common Name TAF6
Gene Symbol TAF6
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LSDIINTPLPRVPLDVCLKAHWLSIEGCQPAIPENPPPAPKEQQKAEATEPLKSAKPGQEEDGPLKGKGQGATTADGKGKEKKAPPLLEGAPLRLKPRSIHELSVEQQLYYKEITEACVGSCEAKRAEALQSIATDPG
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.