missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TAF5L Full-length ORF (NP_001020418.1, 1 a.a. - 325 a.a.) Recombinant Protein with GST-tag at N-terminal

Product Code. 16158542
Change view
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16158542 10 μg 10µg
16168542 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16158542 Supplier Abnova™ Supplier No. H00027097P01.10ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for AP, Array, ELISA, WB-Re

The product of this gene belongs to the WD-repeat TAF5 family of proteins. This gene encodes a protein that is a component of the PCAF histone acetylase complex. The PCAF histone acetylase complex, which is composed of more than 20 polypeptides some of which are TAFs, is required for myogenic transcription and differentiation. TAFs may participate in basal transcription, serve as coactivators, function in promoter recognition or modify general transcription factors to facilitate complex assembly and transcription initiation. The encoded protein is structurally similar to one of the histone-like TAFs, TAF5. Alternatively spliced transcript variants encoding different isoforms have been identified for this gene. [provided by RefSeq]

Sequence: MKRVRTEQIQMAVSCYLKRRQYVDSDGPLKQGLRLSQTAEEMAANLTVQSESGCANIVSAAPCQAEPQQYEVQFGRLRNFLTDSDSQHSHEVMPLLYPLFVYLHLNLVQNSPKSTVESFYSRFHGMFLQNASQKDVIEQLQTTQTIQDILSNFKLRAFLDNKYVVRLQEDSYNYLIRYLQSDNNTALCKVLTLHIHLDVQPAKRTDYQLYASGSSSRSENNGLEPPDMPSPILQNEAALEVLQESIKRVKDGPPSLTTICFYAFYNTEQLLNTAEISPDSKLLAAGFDNSCIKLWSLRSKKLKSEPHQVDVSRIHLACDILEEEV

Specifications

Accession Number NP_001020418.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 27097
Molecular Weight (g/mol) 63.4kDa
Name TAF5L (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MKRVRTEQIQMAVSCYLKRRQYVDSDGPLKQGLRLSQTAEEMAANLTVQSESGCANIVSAAPCQAEPQQYEVQFGRLRNFLTDSDSQHSHEVMPLLYPLFVYLHLNLVQNSPKSTVESFYSRFHGMFLQNASQKDVIEQLQTTQTIQDILSNFKLRAFLDNKYVVRLQEDSYNYLIRYLQSDNNTALCKVLTLHIHLDVQPAKRTDYQLYASGSSSRSENNGLEPPDMPSPILQNEAALEVLQESIKRVKDGPPSLTTICFYAFYNTEQLLNTAEISPDSKLLAAGFDNSCIKLWSLRSKKLKSEPHQVDVSRIHLACDILEEEV
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias PAF65B
Common Name TAF5L
Gene Symbol TAF5L
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.