missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TAF5L Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF6785
This item is not returnable.
View return policy
Description
TAF5L Polyclonal specifically detects TAF5L in Human samples. It is validated for Western Blot.
Specifications
| TAF5L | |
| Polyclonal | |
| Unconjugated | |
| O75529 | |
| TAF5L | |
| E. coli-derived recombinant human TAF5L Met1-Tyr189 Accession # O75529 | |
| 100 μg | |
| Primary | |
| Detects human TAF5L in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human TAF1 is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| PAF65B, PAF65BPAF65-beta, PCAF associated factor 65 beta, PCAF-associated factor 65 beta, TAF5-like RNA polymerase II p300/CBP-associated factor-associated factor 65 kDasubunit 5L, TAF5-like RNA polymerase II, p300/CBP-associated factor (PCAF)-associatedfactor, 65 kD, TAF5-like RNA polymerase II, p300/CBP-associated factor (PCAF)-associatedfactor, 65kDa | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 27097 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction