missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TACE/ADAM17 Ectodomain Antibody, R&D Systems™

Chicken Polyclonal Antibody has been used in 3 publications
Brand: R&D Systems AF930
This item is not returnable.
View return policy
Description
TACE/ADAM17 Polyclonal specifically detects TACE/ADAM17 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| TACE/ADAM17 | |
| Polyclonal | |
| Unconjugated | |
| P78536 | |
| ADAM17 | |
| Insect ovarian cell line T. ni-derived recombinant human TACE/ADAM17 Pro18-Asn671 Accession # P78536 | |
| 100 μg | |
| Primary | |
| Detects human TACE/ADAM17 Ectodomain in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) BACE, rhADAM8, rhADAM10, and recombinant mouse ADAM10 is observed. | |
| Human | |
| IgY |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| ADAM 17, ADAM metallopeptidase domain 17, ADAM metallopeptidase domain 18, ADAM17, ADAM18, CD156b, CD156b antigen, CSVP, disintegrin and metalloproteinase domain-containing protein 17, EC 3.4.24.86, MGC71942, Snake venom-like protease, TACEcSVP, TNF-alpha convertase, TNF-alpha converting enzyme, TNF-alpha-converting enzyme, tumor necrosis factor, alpha, converting enzyme | |
| Chicken | |
| Affinity Purified | |
| RUO | |
| 6868 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction