missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human T-bet/TBX21 Antibody, R&D Systems™
Description
T-bet/TBX21 Monoclonal antibody specifically detects T-bet/TBX21 in Human samples. It is validated for Multiplex Immunofluorescence,Immunohistochemistry
Specifications
Specifications
| Antigen | T-bet/TBX21 |
| Applications | Multiplex Immunofluorescence, Immunohistochemistry |
| Classification | Monoclonal |
| Clone | 3126B |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry 1-10 μg/mL, Multiplex Immunofluorescence 25 μg/mL |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene Accession No. | Q9UL17 |
| Gene Alias | Tbet, T-bet, TBETT-PET, TBLYM, TBLYMT-cell-specific T-box transcription factor T-bet, T-box 21, T-box expressed in T cells, T-box protein 21, T-box transcription factor TBX21, TBX21, T-PET, Transcription factor TBLYM |
| Host Species | Rabbit |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?