missing translation for 'onlineSavingsMsg'
Learn More

Human T-bet/TBX21 Antibody, R&D Systems™

Product Code. 30713212
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30713212 25 μg 25µg
30713211 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30713212 Supplier R&D Systems Supplier No. MAB11692SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

T-bet/TBX21 Monoclonal antibody specifically detects T-bet/TBX21 in Human samples. It is validated for Multiplex Immunofluorescence,Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen T-bet/TBX21
Applications Multiplex Immunofluorescence, Immunohistochemistry
Classification Monoclonal
Clone 3126B
Conjugate Unconjugated
Dilution Immunohistochemistry 1-10 μg/mL, Multiplex Immunofluorescence 25 μg/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene Accession No. Q9UL17
Gene Alias Tbet, T-bet, TBETT-PET, TBLYM, TBLYMT-cell-specific T-box transcription factor T-bet, T-box 21, T-box expressed in T cells, T-box protein 21, T-box transcription factor TBX21, TBX21, T-PET, Transcription factor TBLYM
Host Species Rabbit
Immunogen E. coli derived- recombinant human T-Bet, Glu326-Asp535, Accession # Q9UL17
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30009
Reconstitution Reconstitute lyophilized material at 0.2 mg/ml in sterile PBS. For liquid material, refer to CoA for concentration.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.