missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human T-bet/TBX21 Antibody, R&D Systems™
Mouse Monoclonal Antibody
£188.00 - £467.00
Specifications
| Antigen | T-bet/TBX21 |
|---|---|
| Clone | 1091005 |
| Dilution | Western Blot 0.5 μg/mL, Simple Western 20 μg/mL |
| Applications | Western Blot, Simple Western |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30281238
|
R&D Systems
MAB11637-100 |
100 μg |
£467.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30281239
|
R&D Systems
MAB11637-SP |
25 μg |
£188.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
T-bet/TBX21 Monoclonal antibody specifically detects T-bet/TBX21 in Human samples. It is validated for Western Blot,Simple WesternSpecifications
| T-bet/TBX21 | |
| Western Blot 0.5 μg/mL, Simple Western 20 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. | |
| Tbet, T-bet, TBETT-PET, TBLYM, TBLYMT-cell-specific T-box transcription factor T-bet, T-box 21, T-box expressed in T cells, T-box protein 21, T-box transcription factor TBX21, TBX21, T-PET, Transcription factor TBLYM | |
| E. coli -derived recombinant human T-bet, Glu326-Asp535, Accession # Q9UL17 | |
| Primary | |
| Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. |
| 1091005 | |
| Western Blot, Simple Western | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9UL17 | |
| 30009 | |
| IgG2b | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title