missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human T-bet/TBX21 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems MAB5385-SP
This item is not returnable.
View return policy
Description
T-bet/TBX21 Monoclonal specifically detects T-bet/TBX21 in Human samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, Immunocytochemistry.
Specifications
| T-bet/TBX21 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL | |
| Q9UL17 | |
| TBX21 | |
| E. coli-derived recombinant human T-bet Glu326-Asn535 Accession # Q9UL17 | |
| 25 μg | |
| Primary | |
| Detects human T-bet. | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunocytochemistry | |
| 525803 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Tbet, T-bet, TBETT-PET, TBLYM, TBLYMT-cell-specific T-box transcription factor T-bet, T-box 21, T-box expressed in T cells, T-box protein 21, T-box transcription factor TBX21, TBX21, T-PET, Transcription factor TBLYM | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 30009 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction