missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human T-bet/TBX21 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB53851
This item is not returnable.
View return policy
Description
T-bet/TBX21 Monoclonal specifically detects T-bet/TBX21 in Human samples. It is validated for Intracellular Staining by Flow Cytometry, CyTOF-ready.
Specifications
| T-bet/TBX21 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Tbet, T-bet, TBETT-PET, TBLYM, TBLYMT-cell-specific T-box transcription factor T-bet, T-box 21, T-box expressed in T cells, T-box protein 21, T-box transcription factor TBX21, TBX21, T-PET, Transcription factor TBLYM | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 30009 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Flow Cytometry, CyTOF | |
| 525831 | |
| Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| Q9UL17 | |
| TBX21 | |
| E. coli-derived recombinant human T-bet/TBX21 Glu326-Asn535 Accession # Q9UL17 | |
| 100 μg | |
| Primary | |
| Detects human T-bet/TBX21 in direct ELISAs. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction