missing translation for 'onlineSavingsMsg'
Learn More

Human T-bet/TBX21 Alexa Fluor™ 488-conjugated Antibody, R&D Systems™

Product Code. 30713210
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30713210 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30713210 Supplier R&D Systems Supplier No. IC53851G100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

T-bet/TBX21 Monoclonal antibody specifically detects T-bet/TBX21 in Human samples. It is validated for Flow Cytometry (Intracellular Staining)
TRUSTED_SUSTAINABILITY

Specifications

Antigen T-bet/TBX21
Applications Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone 525831
Conjugate Alexa Fluor 488
Dilution Intracellular Staining by Flow Cytometry 5 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide.
Gene Accession No. Q9UL17
Gene Alias Tbet, T-bet, TBETT-PET, TBLYM, TBLYMT-cell-specific T-box transcription factor T-bet, T-box 21, T-box expressed in T cells, T-box protein 21, T-box transcription factor TBX21, TBX21, T-PET, Transcription factor TBLYM
Host Species Mouse
Immunogen E. coli-derived recombinant human T-bet/TBX21, Glu326-Asn535, Accession # Q9UL17
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30009
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.