missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human SYTL5 Synthetic Peptide
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
SYTL5 Synthetic Peptide, PEP-1290, from Invitrogen.
SYTL5 belongs to the synaptotagmin-like (Slp) protein family, which contains a unique Slp homolog domain (SHD) at the N-terminus. The SHD functions as a binding site for Rab27A, which plays a role in protein transport. Expression of SYTL5 is restricted to placenta and liver, suggesting that it might be involved in Rab27A-dependent membrane trafficking in specific tissues. Several members of the synaptotagmin family are calcium-sensing vesicle-fusion proteins as mediators of cell migration. SYTL7 & SYTL5 were positive regulators of chemotaxis, whereas SYTL2 was a negative regulator of chemotaxis.
Specifications
Specifications
| Gene ID (Entrez) | 94122 |
| Species | Human |
| Purification Method | purified |
| Accession Number | Q8TDW5 |
| Common Name | Human SYTL5 |
| Concentration | 200 μg/mL |
| Content And Storage | -20°C |
| Description | Synthetic Peptide |
| Format | Liquid |
| Formulation | PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction