missing translation for 'onlineSavingsMsg'
Learn More

Human Syntaxin 4 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18778059
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18778059 25 μg 25µg
18122961 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18778059 Supplier R&D Systems Supplier No. MAB7894SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Syntaxin 4 Monoclonal specifically detects Syntaxin 4 in Human samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Syntaxin 4
Applications Western Blot, Immunoprecipitation, Immunocytochemistry
Classification Monoclonal
Clone 884637
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.25 ug/mL, Simple Western 2.5 ug/mL, Immunoprecipitation 2 ug/500 ug cell lysate, Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q12846
Gene Alias NY-REN-31, p35-2, Renal carcinoma antigen NY-REN-31, STX4, STX4Ap35-2, syntaxin 4, syntaxin 4A (placental), syntaxin-4
Gene Symbols STX4
Host Species Mouse
Immunogen E. coli-derived recombinant human Syntaxin 4 Met1-Glu183 Accession # Q12846
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6810
Test Specificity Detects human Syntaxin 4 in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.