missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Syntaxin 4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB7894
This item is not returnable.
View return policy
Description
Syntaxin 4 Monoclonal specifically detects Syntaxin 4 in Human samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.
Specifications
| Syntaxin 4 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.25 ug/mL, Simple Western 2.5 ug/mL, Immunoprecipitation 2 ug/500 ug cell lysate, Immunocytochemistry 8-25 ug/mL | |
| Q12846 | |
| STX4 | |
| E. coli-derived recombinant human Syntaxin 4 Met1-Glu183 Accession # Q12846 | |
| 100 μg | |
| Primary | |
| Detects human Syntaxin 4 in direct ELISAs and Western blots. | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation, Immunocytochemistry | |
| 884637 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| NY-REN-31, p35-2, Renal carcinoma antigen NY-REN-31, STX4, STX4Ap35-2, syntaxin 4, syntaxin 4A (placental), syntaxin-4 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6810 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction