missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Syntaxin 1 (aa 150-177) Control Fragment Recombinant Protein

Product Code. 30195784
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30195784

Brand: Invitrogen™ RP105655

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Syntaxin 1 is a member of the syntaxin superfamily. Syntaxins are nervous system-specific proteins implicated in the docking of synaptic vesicles with the presynaptic plasma membrane. Syntaxins possess a single C-terminal transmembrane domain, a SNARE [Soluble NSF (N-ethylmaleimide-sensitive fusion protein)-Attachment protein REceptor] domain (known as H3), and an N-terminal regulatory domain (Habc). Syntaxins bind synaptotagmin in a calcium-dependent fashion and interact with voltage dependent calcium and potassium channels via the C-terminal H3 domain. This gene product is a key molecule in ion channel regulation and synaptic exocytosis. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q16623
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 6804
Name Human Syntaxin 1 (aa 150-177) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias HPC-1; LOW QUALITY PROTEIN: syntaxin-1 A; neuron-specific antigen HPC-1; P35-1; P35A; Sap; ST x 1; St x 1 A; ST x 1 B; ST x 1B1; ST x 1B2; STXA1; SYN1A; Synaptotagmin-associated 35 kDa protein; syntaxin 1 A; syntaxin 1 A; syntaxin 1 A (brain); syntaxin 1 A (brain)-like; syntaxin-1 A; Syntaxin-1B1; Syntaxin-1B2
Common Name Syntaxin 1
Gene Symbol Stx1a
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence QRQLEITGRTTTSEELEDMLESGNPAIF
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.