missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Syndecan-4 (NP_002990) VersaClone cDNA
Click to view available options
:
10 μg
Description
- Also known as: SYND4
- Syndecan-4 is a transmembrane type I heparan sulfate proteoglycan that functions as a receptor in intracellular signaling.
- It is found as a homodimer and is a member of the syndecan proteoglycan family.
- The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | hSDC4 |
| Accession Number | NP_002990 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 610 bp |
| Gene ID (Entrez) | 6385 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction