missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Syndecan-2/CD362 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF2965-SP
This item is not returnable.
View return policy
Description
Syndecan-2/CD362 Polyclonal specifically detects Syndecan-2/CD362 in Human samples. It is validated for Immunohistochemistry.
Specifications
| Syndecan-2/CD362 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD362, CD362 antigen, Fibroglycan, heparan sulfate proteoglycan 1, cell surface-associated, Heparan sulfate proteoglycan core protein, HSPG, HSPG1, HSPG1syndecan proteoglycan 2, SDC2, SYND2, SYND2cell surface-associated heparan sulfate proteoglycan 1, syndecan 2, syndecan-2, Syndecan2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 6383 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Immunohistochemistry | |
| LYOPH | |
| Immunohistochemistry 5-15 ug/mL | |
| AAH49836 | |
| SDC2 | |
| Mouse myeloma cell line NS0-derived recombinant human Syndecan-2/CD362 Glu19-Glu144 Accession # AAH49836 | |
| 25 μg | |
| Primary | |
| Detects human Syndecan-2/CD362 in direct ELISAs. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) Syndecan-3, rhSyndecan-4, recombinant mouse (rm) Syndecan-1, and rmSyndecan-2 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction