missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SYK (SH2D2 Domain) Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB7166-SP
This item is not returnable.
View return policy
Description
SYK Monoclonal specifically detects SYK in Human samples. It is validated for Western Blot, Simple Western.
Specifications
| SYK | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL, Simple Western 20 ug/mL | |
| P43405 | |
| SYK | |
| E. coli-derived recombinant human SYK (SH2D2 Domain) Trp168-Cys259 Accession # P43405 | |
| 25 μg | |
| Primary | |
| Detects human SYK (SH2D2 Domain) in direct ELISAs. In direct ELISAs, 100% cross-reactivity with the SH2D2 domain of recombinant human (rh) ZAP70 is observed and no cross-reactivity with the SH2 domain of rhCSK is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 720402 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DKFZp313N1010, EC 2.7.10, EC 2.7.10.2, FLJ25043, spleen tyrosine kinaseFLJ37489, tyrosine-protein kinase SYK | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6850 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction